Learn More
CTL Scientific Supply Corp PEHANON INDICATOR STRIPS 1.8-3
Supplier: CTL Scientific Supply Corp 90413
Pehannon Indicator Strips where the indicator and color scale are combined on a single strip. Gradation of scale: 1.8 - 2.1 - 2.4 - 2.7 - 3.0 - 3.2 - 3.5 - 3.8 200 strips/box