Learn More
Perkin Elmer US LLC MEINHARD Concentric Glass Nebulizer, Type C1
Supplier: Perkin Elmer US LLC 00472022
This Type C nebulizer is recommended for high solids applications and features a 1 mL/min sample uptake rate.