Learn More
Biolegend PE/Fire™ 810 anti-human CD8 [SK1]; Isotype: Mouse IgG1 κ; Reactivity: Human Cynomolgus Rhesus; Apps: FC; Size: 25 tests
Supplier: Biolegend 344787
PE/Fire™ 810 anti-human CD8 [SK1]; Isotype: Mouse IgG1 κ; Reactivity: Human Cynomolgus Rhesus; Apps: FC; Size: 25 tests