Learn More
Waters Corp PDA Detector Performance Maintenance Kit
Supplier: Waters Corp WAT052586
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Waters Performance Maintenance Kit for the 996/2996/ACQUITY 2996 PDA Detector contains all of the Waters Quality Parts necessary to keep your instrument operating at peak performance on an annual basis. The Waters Performance Maintenance Kit includes the following parts:a PerformancePLUS Long Life Lamp which is warranted for a lifetime of 2000 hours or 1 year, whichever comes first, and an air filter.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.