Learn More
Cole-Parmer PCRmax Alpha Cycler 2 Thermal Cycler, Dual 96-Well Capacity; 100 to 230 VAC
Supplier: Cole-Parmer 9394522
These compact thermal cyclers deliver reproducible results every runwith little to no training needed. Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification. A Program Wizard generates a protocol specific to your sequence in secondsa quick way to optimize new assays. Units also retain approximately 1000 reports for reviewing at a later date and run on an intuitive, HD Android™ tablet interface. Save your personal settings to your USB drive; sign in is as simple as inserting your USB drive, no password needed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.