Learn More
TWD Scientific LLC Pcr Plate, Twdbiox, 96 Well, Sterile, Polypropylene, Non-Skirted, Printed Axis, Low Profile, 100/Case.
Supplier: TWD Scientific LLC BX603401
BioX 96 Well Clear Polypropylene Non Sterile Non Skirted Low Profile Pcr Plate. Plates Come With Printed Axis. Maximum Capacity Is 200µl. 20 Plates/Pack, 5 Packs/Case, Price/Case
This item is not returnable.
View return policy