Learn More
BrandTech™ BRAND™ PCR 8-Strip Caps
Manufactured from polypropylene without lubricants or chemical additives that can contaminate reagents and samples.PCR 8-Strip Caps are autoclavable and RNase-, DNase-, and DNA-free. Domed caps are available in a variety of colors for sample identification.
Supplier: BrandTech™ BRAND™ 781342
Description
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples.- Available flat or domed
- The domed caps are available in a variety of colors for sample identification
- Autoclavable
- RNase-, DNase-, and DNA-free.
Specifications
Yellow | |
Domed | |
Polypropylene | |
Yes | |
Autoclavable | |
Single-use |
Microplates | |
Non-sterile | |
Yes | |
8-strip Domed Cap | |
Yes | |
Cap Strip |