Learn More
PCB Kit #2, Restek™
Click to view available options
Quantity:
1 mL
Description
- Aroclor standards may exhibit lot-to-lot variation.
- 200μg/mL each in isooctane, 1 mL/ampul
- 32064: Aroclor 1016
- 32065: Aroclor 1221
- 32066: Aroclor 1232
- 32067: Aroclor 1242
- 32068: Aroclor 1248
- 32069: Aroclor 1254
- 32070: Aroclor 1260
Specifications
Concentration | 200μg/mL each in isooctane |
Packaging | Ampul |
Quantity | 1 mL |
Type | Multi-component Standard |
Safety and Handling
ShelfLife | 6 months |