Learn More
Cole-Parmer Oakton PC 450 Waterproof Portable Meter Only
Supplier: Cole-Parmer 3563032
This waterproof meter features intuitive, user-friendly iconselectrode status displays pH slope. Grip-Clip™ electrode holder quickly and easily secures electrode to side of beaker or container (for use with separate pH and conductivity/ATC probes). Memory of 500 data sets and USB or RS-232 output for tranfer to PC. Built-in stand with wall mount allows for easy benchtop measurement or storage.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.