Learn More
Perkin Elmer US LLC PC3-LT Peltier Cooler Organics Sample Introduction Kit for Avio 200/500
Supplier:  Perkin Elmer US LLC N0790025
Encompass_Preferred
Request Bulk Order
The PC3 is a compact Peltier cooled inlet system which incorporates the ESI cyclonic spray chamber. The peltier within the PC3 is air cooled, as such the system can be connected to any ICP. The PC3 cools the outer walls of the cyclonic spray chamber reducing the water or solvent vapor loading on the plasma resulting in enhanced stability and performance. Features and Benefits: Thermal stabilization of spray chamber improves long-term stability Default settings of spray chamber temperature (+2°C/ -5°C) for aqueous or organic solvents. Custom temperature settings are available upon request Interchangeable quartz, PFA and borosilicate cyclonic spray chambers available Fast rinse-out using PFA-ST nebulizer and O-Ring free quartz cyclonic spray chamber Reduced oxides Kit includes: PC3-LT Low temperature Chiller Power Supply Kit Sample Transfer Line Quartz Connector for ZipTorch PFA Connector for Injector ST Sampling Tube 0.15 mm I.D. ST Sampling Tube 0.25 mm I.D. ST Sampling Tube 0.5 mm I
Catalog No. 50-211-3224
Explore available promotions