Learn More
Paulson A-TAC™ “HAWK” Goggles (TN Series)
Click to view available options
Hawk
Description
Includes
Include mounting clips for PASGT (Personnel Armor System, Ground Troops) helmets.
A-TAC Hawk
- Optically correct dual lenses with hard-coated outer lens and anti-fog inner lens
- Black frame fts over most prescription, safety and sun protective spectacles
- Quick-release rear strap buckle
- Noseguard for added protection
- Weighs 5.75 oz.
Compliance
Meet ANSI Z87.1 standards.
Specifications
Certifications/Compliance | ANSI Z87.1 |
Lens Tint | Clear |
Product Type | Tactical Goggle |
Includes | noseguard, mounting clips for PASGT (Personnel Armor System, Ground Troops) helmets |
Frame Material | Silicone |