Learn More
Cole-Parmer Environmental Devices DS-2.5 Ds-2.5:Dust-Sol Pm-2.5 And Pm-10 Dust Sampler
Supplier:  Cole-Parmer 1011190
Encompass
Lightweight and compact, this device uses 47 mm filter media and features a 24 hour timer. It comes with an AC/DC power source with an indicator alerting of a low battery.
Catalog No. 50-630-818