Learn More
Electron Microscopy Sciences QuantomiX WetSEM Particle Imaging Kit
Supplier: Electron Microscopy Sciences PI24
For particles, coating the membrane with a charged polymer attract particles to the membrane. This improve results.