Learn More
Electron Microscopy Sciences Paraffin Wax Paraffin Wax Dispenser
SDP
Supplier:  Electron Microscopy Sciences 6283910
Encompass_Preferred
The Electrothermal Paraffin Wax Dispenser is designed for on-demand delivery of molten wax. It makes routine tissue infiltration and embedding easy and mess free. Designed for use with paraffin wax that has a melting range from 45°C to 65°C. it heats wax to a liquid state and provides convenient "wax on tap" for Histology and Pathology laboratories. The Paraffin Wax Dispenser has a large 7.5 L capacity tank, for melting up to 6 kg of pelletized wax at a time.
Catalog No. 50-243-27