Learn More
Electron Microscopy Sciences Paraffin Wax Dispenser 1 Gallon
Supplier: Electron Microscopy Sciences 6283920
The Electrothermal Paraffin Wax Dispenser is designed for on-demand delivery of molten wax. It makes routine tissue infiltration and embedding easy and mess free. Designed for use with paraffin wax that has a melting range from 45°C to 65°C. it heats wax to a liquid state and provides convenient "wax on tap" for Histology and Pathology laboratories. The Paraffin Wax Dispenser has a large 7.5 L capacity tank, for melting up to 6 kg of pelletized wax at a time.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.