Learn More
Newcomer Supply Paraffin Dispenser XH-4002 with a 2.3 gallon capacity. Digital controller and heats quickly.
Supplier: Newcomer Supply 6527
The Paraffin Dispenser has a 2.3 gallon basin, durable industrial metal outer cabinet, built in temperature controller, stainless steel interior for easy cleaning and heat tolerant metal faucet. It heats quickly and has a Temperature range from room temp to 72°C (tolerance +/- 2°). Overall dimensions: 14" x 16 1/4" x 19". Interior size: 9" diameter x 10". CE certified and cord is CSA & UL listed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.