Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Panexin basic, Serum Replacement with Defined Components (100 ml)
Supplier: ILEX LIFE SCIENCES LLC P0496900
Panexin basic, Serum Replacement with Defined Components. QUANTITY: 100 ml. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: -20 degrees Celsius. DESCRIPTION: Panexin basic is a fully defined serum replacement for the cultivation of adherent and non-adherent cells under serum-free culture conditions or to significantly reduce the amount of serum in cell culture. It supports the growth of many cell types in an optimum manner without any extra handling compared to serum. COMPOSITION: Contains purified proteins (less than 1 % w/v), lipids, salts, amino acids, trace elements, hormones and only traces of animal-derived components (less than 1 % w/v). It contains no growth factors, undefined hydrolysates or peptones.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.