Learn More
Genscript Corporation Streptavidin Antibody, pAb, Rabbit
Supplier:  Genscript Corporation A006215
Encompass_Preferred
GenScript Streptavidin Antibody is specific to streptavidin. Streptavidin is a tetrameric protein from Streptomyces avidinii that binds very tightly to the vitamin biotin with a Kd of ≈10-14 mol/L.The high-affinity recognition of biotin and biotinylated molecules has made streptavidin one of the most important components in diagnostics and laboratory kits.GenScript Streptavidin Antibody is developed in rabbit using purified Streptavidin and highly purified from rabbit antiserum by protein G resin.
Catalog No. 50-272-852
Explore available promotions