Learn More
AUTOMATION TRAINER P30XL, Clear, Filtered, Sterile, Racked (50 384-Tip Racks/Case)
Supplier: AUTOMATION TRAINER BP30XLCLFSR384
This item has a minimum order quantity of 10 per supplier requirements.
Automation Trainer's Beckman-Compatible 384-Format Pipette Tips are designed to be a direct replacement to Beckman's 384-Format P30 & P30XL tip. Our tips are manufactured to strict standards to ensure quality and consistent performance. Automation control software protocol modification and separate labware definition is not required. Maximum working volumes are 30 L for the Unfiltered P30 Tip and 50 L for the Unfiltered P30XL Tip.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.