Learn More
Honeywell™ Combination Organic Vapor Cartridge and HEPA Filter Cartridge
Designed for use with Honeywell North Primair 700 Series Powered Air Purifying Respirator (PAPR).
Supplier: Honeywell™ PA7OVHE
This item is not returnable.
View return policy
Description
- Filter locks into place with a quarter turn, assuring workers they are secure
- HEPA filter provides 99.97% minimum filter efficiency for all particulates
Compliance
All North filters are NIOSH approved certified to 42CFR84 standards when used with North respirators.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
HEPA, Organic Vapor | |
Threaded | |
Combination Cartridge/Filter | |
PA700 |
NIOSH Approved | |
Black/Magenta | |
Primair 700 Series PAPR | |
4/Pk. |