Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonic Acid Monohydrate, Certified ACS, Fisher Chemical

Catalog Number A320500
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
6192-52-5
C7H10O4S
190.213
MFCD00142137
KJIFKLIQANRMOU-UHFFFAOYSA-N
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
521998
4-methylbenzenesulfonic acid;hydrate
CC1=CC=C(C=C1)S(=O)(=O)O.O
6192-52-5
C7H10O4S
190.213
MFCD00142137
KJIFKLIQANRMOU-UHFFFAOYSA-N
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
521998
4-methylbenzenesulfonic acid;hydrate
CC1=CC=C(C=C1)S(=O)(=O)O.O
Melting Point 100°C
Boiling Point 140°C (20 mmHg)
Color White
Linear Formula CH3C6H4SO3H·H2O
UN Number 2585
Formula Weight 190.22
Grade Certified ACS
Quantity 500 g
Packaging Glass Bottle
Physical Form Solid
Chemical Name or Material p-Toluenesulfonic Acid Monohydrate
Show More Show Less
DOTInformation Class 8: Corrosive
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit