Learn More
Thermo Scientific™ Oxoid™ TST-RPLA Kit
Oxoid TST-RPLA Kit detects staphylococcal toxic-shock syndrome toxin (TST) by reversed passive latex agglutination.
Supplier: Thermo Scientific™ TD0940A
Description
Oxoid TST-RPLA Kit detects staphylococcal toxic-shock syndrome toxin (TST) by reversed passive latex agglutination.
Specifications
Staphylococcus | |
Microbiology Media | |
20 Tests/Kit |
2°C to 8°C | |
Reverse Passive Latex Agglutination | |
Staphylococcal Toxic-Shock Syndrome Toxin (TST) |