Learn More
Thermo Scientific™ Biochemical Identification System (O.B.I.S.) Mono
Rapidly differentiate Listeria monocytogenes from other Listeria species with this biochemical identification System.
Supplier: Thermo Scientific™ ID0600M
Description
Rapidly differentiate Listeria monocytogenes from other Listeria species with Thermo Scientific™ Oxoid™ Biochemical Identification System (O.B.I.S.) mono. This rapid colorimetric test delivers simple and rapid results for the determination of D-alanyl aminopeptidase (DALAase). It has been designed for the differentiation of presumptive Listeria species that have been isolated from selective media and plated onto a secondary medium for further biochemical testing.
Identification of Listeria monocytogenes relies on the lack of the enzyme D-alanyl aminopeptidase.1,2,3
Listeria monocytogenes is the most common pathogenic Listeria spp. and has been shown to be pathogenic to both humans and animals.
Use O.B.I.S. mono to rapidly differentiate Listeria monocytogenes from other Listeria spp., reducing the need for a full biochemical identification of suspect colonies.
- Simple—easy to implement
- Rapid—results in 6 minutes of testing suspect colonies
- Non-carcinogenic substrate
We have developed a new system for aminopeptidase testing4 using a non-carcinogenic substrate in response to health concerns associated with amino acid conjugates of ß naphthylamine5,6 as these are potent carcinogens.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
L. monocytogenes | |
60 Tests/Kit |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.