Learn More
Thermo Scientific™ Oxoid™ Microbact™ Listeria 12L Kit
Simplify the identification of Listeria spp., including Listeria monocytogenes, with the Thermo Scientific™ Oxoid™ Microbact™ Listeria 12L Kit.
Supplier: Thermo Scientific™ MB1128A
Description
Simplify the identification of Listeria spp., including Listeria monocytogenes, the principal causative agent of human listeriosis, with the Thermo Scientific™ Oxoid™ Microbact™ Listeria 12L Kit—a complete, self-contained biochemical-based identification system. Each strip incorporates 12 analytes, 11 carbohydrate utilization tests and one rapid microhaemolysis test, and allows for fast, reliable and easy-to-read results.
Listeria spp. are widespread in the environment and have been isolated from many different foods and animals. In severe cases, it can cause septicemia, meningitis, encephalitis and, in pregnant women, intrauterine infections that may lead to spontaneous abortion or stillbirth. With this kit, Listeria spp. are quickly and accurately identified so that proper, fast treatment for the patient may follow.- Fast: Accurate identifications in as little as four hours
- Simple: No prior subculturing on blood agar, no CAMP test and no developing reagents required
- Easy-to-Read and Interpret: Distinct color changes and settling pattern are read visually and interpreted easily using the Microbact Identification Package (MB1244A)
- Comprehensive: Allows the definitive identification of all Listeria spp., including L. monocytogenes and L. innocua.
- Flexible: Ideal for use with chromogenic or selective media, in clinical or food laboratory settings
- Reliable: Substrates include all those recommended in recognized international standard methods, including FDA BAM and ISO
Microbact users: Download Thermo Scientific™ Microbact™ Software and stay up-to-date with our latest updates by visiting our Biochemical Identification Software Downloads portal >
Not all products are available for sale in all territories. Please inquire.
Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Microbact Listeria 12L Kit | |
20 test strips and suspending medium vials (3mL), holding tray, technical product insert and report forms | |
20 Tests/Kit |
Listeria species | |
1 Box |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.