Learn More
Thermo Scientific™ Oxoid™ IMAGEN™ Respiratory Screen Positive and Negative Control Slide
Slide with 1 row of wells containing separate preparations from cell cultures infected with RSV, influenza A and B virus, parainfluenza virus 1, 2 and 3 or adenovirus.
Supplier: Thermo Scientific™ S6121302
Description
The IMAGEN™ Control Slides are for use as a control to the following tests: IMAGEN Adenovirus (K610011-2) IMAGEN Respiratory Syncyti al Virus (RSV) (K610211-2) IMAGEN Parainfluenza Group (K610311-2) IMAGEN Parainfluenza Typing 1, 2 and 3 (K610411-2) IMAGEN Influenza A and B Virus (K610511-2) IMAGEN Respiratory Screen (K612011-2) IMAGEN hMPV (K612511-2) IMAGEN Herpes Simplex Virus (HSV) (K610611-2) IMAGEN Chlamydia kit (K610111-2)
The second row of wells contains preparations from uninfected cell cultures for use as negative controls. Sealed in foil pouch filled with nitrogen.Specifications
IMAGEN™ Respiratory Screen Positive and Negative Control Slide | |
Control | |
1 Slide/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.