Learn More
Thermo Scientific™ Oxoid™ Dip-Slides™ Industrial
Make colony counting simple for aerobic bacteria, yeast, mold, coliform bacteria and Enterobacteriaceae with the Thermo Scientific™ Oxoid™ Dip-Slides™ Industrial.
$88.00 - $92.00
Specifications
Content And Storage | 15°C to 25°C |
---|---|
Format | Dip Slide |
Product Type | Sample Collection |
Quantity | 10/Pk. |
Catalog Number | Mfr. No. | Description | dimFormat | Quantity | Manufacturing Origin | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | dimFormat | Quantity | Manufacturing Origin | Price | Quantity & Availability | ||
DS0147A
|
Thermo Scientific™
DS0147A |
Dip Slide TTC (Red Spot) | 10/Pk. | United Kingdom |
Pack of 10 for $88.00
|
|
|||
DS0155A
|
Thermo Scientific™
DS0155A |
Dip Slide TTC (Red Spot) and Malt Extract Agar | 10/Pk. | United Kingdom |
Pack of 10 for $92.00
|
|
|||
Description
Make colony counting simple for aerobic bacteria, yeast, mold, coliform bacteria and Enterobacteriaceae with the easily disposable Thermo Scientific™ Oxoid™ Dip-Slides™ Industrial. Simple-to-use for the operator and where laboratory facilities are limited, the Dip Slide is coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective.
Originally designed to determine bacterial counts on urine specimens, dip slides have been modified over the years by making the device double-sided, with one side a medium for Gram positive species and the other side a medium for Gram negatives. With these dip slides, the process for colony counting is time- and cost-effective, allowing exposure of the specimens to two different media simultaneously.- Slide is made of easily disposable plastic with raised edges to ensure an even thickness of culture medium
- Molded grid allows for easier colony counting
- Slide is shaped to ensure drainage of fluid from the slide
- Cap forms a convenient handle by which it may be held without risk of touching the culture medium
- Bacteria, fungal and yeast charts are included, make examining growth simple for the operator and where laboratory facilities are limited
Specifications
15°C to 25°C | |
Dip Slide | |
Sample Collection | |
10/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.