Learn More
Trinity Biotech, Inc OXALATE REAGENTS /QUANTITATIVE
Supplier:  Trinity Biotech, Inc 591D1KT
Encompass_Preferred
Trinity Biotech Oxalate reagents are for the quantitative, enzymatic determination of oxalate in urine at 590 nm
Catalog No. NC2224364