Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Owl™ JGC Gel Casting Systems
Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.
Supplier: Thermo Scientific™ JGC4
Description
Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.
- Pouch collects excess polyacrylamide and seals bottom of gel, eliminating need for tape or bottom spacers.
- Design allows storage of gels without dehydration by adding buffer and heat sealing.
- System casts from one to four gels.
- Few moving parts result in outstanding durability, extra long life.
Includes:
- Casting stand
- Blocking plate
- Three-point leveling base
- Spacer placer
- 25 gel pouches
Specifications
Gel Electrophoresis | |
16 x 14 cm | |
Gel Caster | |
5.13 in. | |
10 cm | |
13 cm |
Owl JGC Gel Casting System | |
Owl | |
10 cm | |
13 cm | |
5.13 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction