Learn More
Chemglass Life Sciences 5 Cu. Ft. Oven, Forced Air Convection, Bench Top, Multi-Purpose, 120V
Supplier: Chemglass Life Sciences CLS344605F
5 Cu. Ft. Oven, Forced Air Convection, Bench Top, Multi-Purpose, 120V. The new multi-purpose ovens feature microprocessor controls & digital temperature display providing optimal stability and allows for accurate control. The set-points have memory capabilities in the event of power outages.The digital timer provides the user w/ the ability to automatically turn off the oven at the end of an application cycle. The timer has a range of one minute to 99 hours/59 minutes.The model's safety controller is an over-temperature protection (OTP) that operates independently from the main controller. The dual-control, failsafe function means a second system will ensure that temperature set-point is never exceeded.The bottom of the chamber is solid & separated from the heating elements. Heating elements are non-contact & low watt density. Applications include: glassware drying, sterilizing, aging testing, stability testing & electronic burn-in. 24 month warranty.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.