Learn More
Novus Biologicals™ Ovary Tissue Slides - Paraffin
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Novus Biologicals™ NBP230294
Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.Specifications
Human | |
Ovary Tissue Slides | |
This product is for research use only and is not approved for use in humans or in clinical diagnosis. | |
Human |
Safety and Handling
Recommended Storage : Store at 4°C. Do not freeze.