Learn More
Electron Microscopy Sciences EMS Osteotome U.S. Army Premium, 18 mm, 6½" OAL
Supplier:  Electron Microscopy Sciences 7799014
Encompass_Preferred
These U.S. Army pattern osteotomes, also called bone chisels, are used to score, cut, scrape, clean and sculpt the bone.
Size: 18 mm
OAL: 6½"
Catalog No. 50-193-1577