Learn More
Electron Microscopy Sciences EMS Osteotome Lambotte Standard, 1" edge, 8" OAL
Supplier: Electron Microscopy Sciences 7799016
These osteotomes feature the Lambotte pattern and are used to score, cut, scrape, clean and sculpt the bone.
Cutting edge: 1"
OAL: 8"