Learn More
Electron Microscopy Sciences EMS Osteotome Lambotte Premium, 1" edge, 9" OAL
Supplier: Electron Microscopy Sciences 7799018
These osteotomes feature the Lambotte pattern and are used to score, cut, scrape, clean and sculpt the bone.
Cutting edge: 1"
OAL: 9"