Learn More
Moc Incorporated OSRAM XENO 6VOLT 20WATT 5/PK
Supplier: Moc Incorporated HLX64250
Osram Sylvania HLX 64250 (FHE/ESB) 20 Watt 6 Volt Halogen Lamp. Specifications: ANSI Code: FHE/ESB; Wattage: 20 Watts; Voltage: 6 Volts; Filament: C-6 Tungsten; Length (MOL): 1.2 IN; Base: 2-PIN (G4); Shape: 3.5 Initial Light Lumens: 480 lm; Average Bulb Life: 100 Hours. 5/pack.