Learn More
TEKTRONIX INC OSCILLOSCOPE,200MHZ,1GS/S,BASE
Supplier: TEKTRONIX INC TBS1202C
Digital Storage Oscilloscope: 200 MHz bandwidth, 1 GS/s sample rate, 2 Channel, 20Kpts record length*Base unit only, No accessories*
This item is not returnable.
View return policy