Learn More
PATTERSON DENTAL COMPANY ORTHOJET LIQUID CLR,8OZ,1304-C
Supplier:  PATTERSON DENTAL COMPANY 4594412
Encompass_Preferred
Ortho-Jet Liquid, Clear, 8 ozLang Dental Manufacturing Co IncOrtho-Jet is a fast, self-curing acrylic resin with extra clarity for orthodontic appliances. This product is intended for the fabrication of orthodontic appliances.Extra-fine bead size – Adapts to the model better for an accurate, comfortable fitCross-linking monomer – Gives the acrylic strength and durabilityLow flow properties – Allows better control and will not slump, with minimal shrinkingSuperior clarity – Creates an aesthetically pleasing applianceAdditional uses – Perfect for occlusal splints and nightguards and resists wear
Catalog No. NC2244091