Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Testosterone
Supplier: Ortho-Clinical Diagnostics™ 1435205
This item is not returnable.
View return policy
Specifications
Testosterone | |
Vitros™ Testosterone Reagent | |
100 Pack | |
32 min. |
Well | |
Coated wells 100, conjugate reagent 8.4mL, Biotinylated Antibody reagent 13.3mL | |
25 μL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction