Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Calibrators: Hepatitis B Surface Antigen
Supplier: Ortho-Clinical Diagnostics™ 6801323
This item is not returnable.
View return policy
Specifications
2°C to 8°C | |
VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system | |
Hepatitis B Surface Antigen (HBSAG) Calibrator | |
Qualitative in-vitro detection of Hepatitis B Surface Antigen (HBsAg) |
Vial | |
Human Serum and Plasma | |
2mL VITROS™ HBsAg calibrator (Human HBsAg ad subtype, inactivated in Buffer with Bovine Serum Albumin and antimicrobial agent, lot calibration card, protocol card, 8 calibrator bar code labels | |
1 Vial/Pk. |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction