Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Calibrators: Folate
Supplier: Ortho-Clinical Diagnostics™ 1311075
This item is not returnable.
View return policy
Specifications
2°C to 8°C | |
Human Serum, Plasma (Heparin), Whole Blood | |
2.1mL VITROS™ Folate calibrators 1, 2 and 3 (Folic Acid in Buffer with Human Serum Albumin and antimicrobial agent), lot calibration card (for use with reagent pack 1, colored green), ancillary lot card (for use with reagent pack 2, colo | |
3 |
VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system | |
Folate Calibrator | |
Quantitative measurement of Folate | |
1 Set/Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction