Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Calibrators: Estradiol
Supplier: Ortho-Clinical Diagnostics™ 1318930
This item is not returnable.
View return policy
Specifications
2°C to 8°C | |
Human Serum and Plasma (EDTA or Heparin) | |
2mL VITROS™ Estradiol calibrators 1 and 2 (Estradiol in Human Serum, with antimicrobial agent), lot calibration card, protocol card, 16 calibrator bar code labels (8 for each calibrator) | |
2 |
VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system | |
Estradiol Calibrator | |
Quantitative measurement of Estradiol | |
1 Set/Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction