Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORP Standard, 400 mV, Spectrum™ Chemical
Supplier: Spectrum Chemical Manufacturing Corporation O212500MLGL
Specifications
7447-40-7, 14459-95-1, 13746-66-2 | |
Amber Glass Bottle | |
500 mL |
Chemical Solution | |
Yellow |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Spot an opportunity for improvement?
Provide Content Correction