Learn More
Sensorex ORP Sensor,Gold,Ultem,DJ,BNC
Supplier: Sensorex ORP2000AU
The ORP2000 with gold band features double junction reference protection, which prevents contamination, by sample components that interact with silver (Ag). The durable Ultem body ensures extensive chemical compatibility.