Learn More
Sensorex ORP Sensor, Gold, SJ,BNC
Supplier: Sensorex ORP1000AU
The ORP1000 with gold band is a durable and easy to use ORP sensor and has built in protection for the ORP sensor surface. Use with any ORP meter accepting a BNC connector.