Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Orion™ Star A211 pH Benchtop Meter Kit PROMO
Reliably measure pH, mV, ORP and temperature with the Thermo Scientific™ Orion™ Star A211 pH Benchtop Meter for advanced lab analysis and printer or computer interfacing
Supplier: Thermo Scientific™ STARA2118PM
Description
Thermo Scientific Orion Star A211 pH benchtop meters are easy to use, dependable laboratory meters with advanced features and functions. Selectable read modes and stability indicator give you flexibility and control over your testing. All critical data is shown with your measurements including temperature, date and time, read type, measurement stability, active calibration information and electrode condition icon. Onscreen calibration and setup instructions can be displayed in several local languages with new languages added through software updates.Navigation is fast and simple due to the comprehensive keypad with menu-specific function keys, dual purpose scroll/shortcut keys, plus power, measure, log view and stirrer keys that ensure easy operation. All setup menus are in one location with direct access using a single shortcut key so you can quickly access, evaluate and change measurement and instrument settings.
- Measure pH, mV, relative mV or ORP with temperature
- Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
- Calibration editing option allows individual points to be fixed without a full recalibration
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Capture your critical readings with measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
- Eliminate the need for magnetic stir bars using the meter-controlled stirrer probe (catalog number 096019) to uniformly mix solutions
- Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
- Calibration log stores ten most recent calibrations
- Non-volatile meter memory ensures your data and settings are protected, even with loss of power
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
- Flexible power options with universal power adapter or optional AA batteries
- IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited
- Meters include CE, TUV 3-in-1 and FCC Class A certifications and 3-year replacement warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Orion Star A211 pH benchtop meter kit with ROSS Ultra pH electrode, ATC probe, solutions and stand | |
±0.1°C | |
±0.2 or ±0.05%, whichever is greater | |
Star A211 pH meter, 8102BNUWP ROSS Ultra glass-body pH electrode, 927007MD stainless steel ATC probe, 810199 ROSS pH buffer and solution kit, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and | |
pH with calibration editing option, relative mV, ORP and temperature | |
0.001, 0.01, 0.1 | |
±2000.0 mV | |
1 to 5 | |
22.0°F to 221.0°F | |
Manual or automatic with ATC temperature probe | |
LCD, Graphic with Backlight | |
Non-volatile memory preserves data log, calibration log and meter settings | |
Automatic with Auto-Read and Timed Read Modes, Manual with Continuous Read Mode | |
BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe) | |
IP-54 | |
24 x 18 x 11 cm (9.5 x 7.1 x 4.3 in.) | |
2 lb. | |
10 per channel with password protection | |
Universal AC adapter (included) or 4 AA batteries (sold separately) | |
English, Spanish, French, Italian, German and Chinese; Portuguese and Korean Available with Software Update | |
5 to 85 %, non-condensing | |
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting | |
5°C to 45°C (ambient) | |
4 x AA (sold separately) |
Transfer single, range or all data points to printer or computer | |
±0.002 | |
USB, RS-232 | |
pH, mV, Relative mV, ORP with Temperature | |
-2.000 to 20.000 | |
±2000 | |
0.1 mV | |
-5.0°C to +105.0°C | |
0.1°C, 0.1°F | |
1 point temperature offset calibration | |
Yes, selectable | |
2000 with Date and Time Stamp | |
High/low limit alarm, calibration due alarm | |
CE, TUV 3-1, FCC Class A | |
Universal 50 to 60 Hz, 100-240 VAC power adapter (Cat. No. 1010003) | |
0.9 kg | |
Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys | |
Orion Star A211 pH/mV/RmV/ORP/Temperature Benchtop Meter | |
pH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe, stirrer probe (includes 8102BNUWP ROSS Ultra pH electrode and 927007MD ATC probe) | |
±2000.0 mV | |
7 pH | |
41°F to 113°F (ambient) | |
1 | |
800 hr. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction