Learn More
Thermo Scientific™ Orion™ Star A111 Benchtop pH Meter Kit PROMO
Take simple, accurate pH measurements with this easy-to-use, affordable Thermo Scientific™ Orion Star™ A111 Benchtop pH Meter.
Supplier: Thermo Scientific™ STARA1117PM
Description
Features:- Large, informative screen clearly shows the main information, including pH or mV reading, temperature, meter mode, calibration information and power source.
- Thermo Scientific™ AUTO-READ™ locks in the stable reading on your screen and ready indicator alerts when readings are stable
- Meter offers up to three point pH calibration and automatic recognition of USA/NIST and DIN buffers.
- Use can easily recall calibration slope data for procedure checks.
- Nonvolatile memory holds up to 50 data points.
- Included electrode arm and newly designed probe holder make it easier to maintain and place probes into samples.
- Plug the meter into almost any power outlet with the included universal power adapter or use four AA batteries (sold separately) to run the meter on DC power.
- The IP54-rated housing can handle splashes and can be wall-mounted if table space is a problem.
Orion STAR: A111 Benchtop pH Meter, Part no. STARA1110, includes electrode arm with redesigned holder and universal power adapter
Orion STAR: A111 Benchtop pH Meter Kit, Part no. STARA1115, includes 9157BNUMD Orion Triode™ 3-in-1 pH/ATC probe; Solution kit (60mL each of pH 4, 7 and 10 buffers; storage solution and cleaning solution); electrode arm with redesigned holder and universal power adapter
Orion STAR: A111 Benchtop pH Meter Kit, Part no. STARA1116, includes GD9156BNWP Orion Green electrode; 927007MD Stainless Steel ATC Probe; electrode arm with redesigned holder and universal power adapter
Specifications
Orion Star A111 pH benchtop meter kit with ROSS pH/ATC electrode, solutions and stand | |
±0.2 or ±0.05%, whichever is greater | |
Internal Data Point Memory | |
Star A111 pH meter, 8157BNUMD ROSS Ultra Triode epoxy-body pH/ATC electrode, 810199 ROSS pH buffer and solution kit, electrode stand, universal power adapter, literature CD, printed quick start guide and meter test certificate | |
pH, relative mV (RmV) and temperature | |
0.01, 0.1 | |
0.1 mV | |
-5.0°C to +105.0°C | |
0.1°C, 0.1°F | |
1 point temperature offset calibration | |
Non-volatile memory preserves log and settings | |
Automatic with Auto-Read Measure Mode, Manual with Continuous Measure Node | |
CE, TUV 3-1, FCC Class A | |
Universal 50 to 60 Hz, 100-240 VAC power adapter (Cat. No. 1010003) | |
0.9 kg | |
3 Year Meter Warranty | |
Orion Star A111 pH/mV/RmV/Temperature Benchtop Meter | |
pH electrode, ORP electrode, ATC temperature probe (includes 8157BNUMD ROSS Ultra Triode pH/ATC electrode) | |
±1999.9 mV | |
7 pH | |
41°F to 113°F (ambient) | |
1 | |
2000 hr. |
±0.1°C | |
±0.01 | |
None | |
pH, mV, Relative mV with Temperature | |
-2.00 to 16.00 | |
±1600.0 | |
1 to 3 | |
22.0°F to 221.0°F | |
Manual or automatic with ATC temperature probe | |
LCD | |
50 | |
BNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe) | |
IP-54 | |
24 x 18 x 11 cm (9.5 x 7.1 x 4.3 in.) | |
2 lb. | |
Simple with highlighted measure key, scroll and shortcut keys | |
Universal AC adapter (included) or 4 AA batteries (sold separately) | |
English | |
5 to 85 %, non-condensing | |
Smart Stability and Smart Averaging | |
5°C to 45°C (ambient) | |
4 x AA (sold separately) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.