Learn More
Orion™ pH 4.01 Buffer, Individual Use, Pink, 25 Pouches, pH 4.01 Buffer, Color Coded Pink, 25 Pouches PROMO, Thermo Scientific™
Ensure fresh, ready to use standards for pH calibration with Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches.
Supplier: Thermo Scientific 910425PM
Description
- Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
- Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
- Manufactured under ISO 9000 quality standards
- Pouches are 15mL each and are available in 10 packs or 25 packs
Note:
The following part numbers have been changed due to a recent system upgrade.
Old Part Number: 910410
New Part Number: 910410-WA
Old Part Number: 911025
New Part Number: 911025-WA
Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Data Safety Sheet (MSDS) files.
Specifications
pH Buffer, individual use pouches | |
pH calibration | |
4.01 | |
Buffer | |
Yes |
pH 4.01 Buffer | |
pH Meter and Electrode, single-use | |
25 x 15 mL | |
Pouch |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.