Learn More
Thermo Scientific™ Orion™ Dual Star™ pH, ISE, mV, ORP and Temperature Dual Channel Benchtop Meter Kit PROMO
Accurately monitor pH, ion concentration, mV, ORP and temperature on two channels with the Thermo Scientific™ Orion™ Dual Star pH / ISE Benchtop Meter for advanced lab analysis.
Supplier: Thermo Scientific™ 2115201PM
Description
The Orion Dual Star pH/ISE dual channel benchtop meter accelerates sample measurement throughput by showing the results of two channels simultaneously on its large, backlit display. Measure pH, ion concentration (ISE), mV, relative mV or ORP with temperature on two channels using the meter’s two BNC, reference pin-tip and ATC temperature inputs. Collect critical measurements using an assortment of read modes and onscreen ready indicator that alerts when readings are stable. The meter’s Smart Stability and Smart Averaging functions automatically compensate for measurement conditions, so the meter displays more accurate results faster.
- View large individual measurements on a full screen display or quickly toggle to a split screen display to view two measurements at the same time
- Easy-to-follow onscreen text prompts for calibration and setup menus available in a multi-language user interface with English, Spanish, French, German or Italian options
- EZ Startup menu facilitates quick and easy first time meter setup
- Numeric keypad and menu-specific function keys allow for quick and efficient meter setup and data entry
- Selectable read modes include Auto-Read, On-Ready, At Timed Intervals, Continuous, Value Change in Measurement and Timed for a variety of measurement collection methods for different analytical methods
- Perform up to a 6 point pH calibration with automatic recognition for USA/NIST and EURO/DIN buffers with option to manually input custom buffer values
- Perform up to a 6 point ion concentration (ISE) calibration with non-linear auto-blank and low-concentration stability options or use incremental methods for single/double known addition and single/double known subtraction
- Non-volatile meter memory holds up to 1000 data points with time and date stamp
- Easily transfer data via USB and RS232 to a printer or computer with free Orion Dual Star Com communication software
- Stir samples with ease using the meter-controlled Orion Star stirrer probe
- Meter-attachable electrode stand assists with moving and maintaining electrodes
- IP54-rated housing handles splashes and is wall-mountable to save space
Warranty: Three-year meter replacement warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Universal 50 to 60 Hz, 100 to 240 VAC power adapter (Cat. No. 1010003) | |
USB, RS232 | |
±0.002 | |
Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit | |
Yes, adjustable | |
1 point relative mV (RmV) mode, 1 point ORP mode | |
CE, TUV 3-1, FCC Class A | |
1000 with date and time stamp | |
2 to 6 | |
pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature | |
±1999.9 mV | |
0.0001 minimum, 1 to 3 significant digits (user selectable) | |
-5.0°C to +105.0°C | |
Dual Star pH/ISE dual channel meter, 9512BNWP Orion ammonia ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 951207 Orion 100 ppm ammonia standard (475 mL), 951211 Orion ammonia ISA ionic strength adjuster (475 mL), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate | |
7 pH (adjustable), 1.000 ISE (adjustable) | |
English, Spanish, French, Italian and German | |
Non-volatile memory preserves data log, calibration log and meter settings | |
Orion Dual Star pH/ISE/mV/RmV/ORP/Temperature Dual Channel Benchtop Meter | |
pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe, stirrer probe (includes 8102BNUWP ROSS Ultra pH electrode, 927007MD Orion ATC probe) | |
±1999.9 mV | |
0.1 mV | |
Smart Stability and Smart Averaging | |
5°C to 45°C (ambient) | |
Manual or automatic with ATC temperature probe | |
24 x 17 x 9 cm (9.5 x 6.7 x 3.5 in.) | |
0.9 kg |
±0.2 mV or ±0.05% of reading whichever is greater | |
±0.2 mV or ±0.05% of reading whichever is greater | |
±0.1°C | |
High/low limit alarm | |
pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ | |
1 to 6 | |
Transfer single, range or all data points to printer or computer | |
Orion Dual Star pH/ISE dual channel benchtop meter ammonia and pH kit with Orion ammonia ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and stand | |
2 | |
0.0001 to 19,900 in ppm, M, mg/L, % or unit-less | |
-2.000 to 19.999 | |
0.1, 0.01, 0.001 | |
Graphic LCD with Backlight | |
Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe) | |
Numeric with menu-specific function keys and dual purpose scroll/shortcut keys | |
Automatic data logging with Auto-Read, On Ready, At Timed Intervals, Value Change In Measurement and | |
10 per channel with password protection | |
Universal AC adapter (included) | |
±1999.9 mV | |
5 to 85 %, non-condensing | |
0.1°C, 0.1°F | |
41°F to 113°F (ambient) | |
1 point temperature offset calibration | |
3 Year Meter Warranty | |
2 lb. | |
90 to 260 VAC 50/60 Hz |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.