Learn More
Thermo Scientific™ Orion™ AquaMate 7000 Vis Spectrophotometer
Choose the accuracy, dependability and ease of use of the Thermo Scientific™ Orion™ AquaMate 7000 Vis Spectrophotometer to optimize your water and wastewater testing
Supplier: Thermo Scientific™ AQ7000
Description
Choose the accuracy, dependability and ease of use of the Thermo Scientific™ Orion™ AquaMate 7000 Vis Spectrophotometer to optimize your water and wastewater testing. This dependable lab spectrophotometer includes 260 preprogrammed methods for ion concentration from absorbance or create custom methods using the standard curve function so virtually any brand of colorimetric reagent can be used. Utilize 325-1100 nm selectable wavelengths, 5 nm spectral bandwidth and vial size flexibility with the included 3-position carousel to hold 13-24 mm round, 10 mm square and 20-50 mm rectangular vials.
Pre-programmed Methods:
- More than 260 preprogrammed methods offers easy and convenient measurements using Orion AQUAfast™, Merck™ or CHEMetrics™ chemistries
- Preprogrammed methods can also be adapted for new chemistries or operators can create their own custom methods and calibration curves
- Instruments allow a one-point adjustment on any preprogrammed method to correct for variations in batch reagent chemistries
Versatility:
- Wide variety of vials sizes can be used, including 10mm square vials, 20 and 50mm rectangular vials and an adjustable round vial holder that accommodates 13, 16, and 24mm round vials
- Three USB connections provide the options to store methods and data to a USB memory stick, transfer data to a computer and print hard copy data to an external printer
- System is compatible with ink jet and laser printers with HP PCL 5 or greater
Verification:
- Performance verification tests ensure wavelength accuracy and instrument functionality
- Each verification report gives the time, date and instrument serial number in accordance with GLP and GMP
- Built-in wavelength accuracy test is compatible with either the internal lamp or external calibrated standards
- Built-in test methods for stray light, noise and resolution are available to validate instrument performance further
Includes:
- Methods on USB memory stick
- Three-position cell holder
- User documentation
- Dust cover
- USB cable
- 110V, 220V and 240V power cords
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Compatible with ink jet and laser printers with HP PCL 5 or greater | |
AquaMate 7000 Vis Spectrophotometer | |
Graphical with LCD backlight, 9.7 x 7.1 cm (3.8 x 2.8 in.) | |
5 nm | |
325 to 1100 nm | |
100/240 V, 50/60 Hz | |
25 cm | |
Methods on USB memory stick; 3-position cell older; User documentation; Dust cover; USB cable 110V and 220V power cords | |
1000 hrs. | |
Single Beam | |
-0.1 to 3.0 A ; -0.3 to 125 %T ; ±9999 C | |
10 to 4200 nm/min. | |
±1 nm | |
±0.5nm | |
19 lb. | |
11.8 in. |
USB type A port for USB stick (front panel) USB type B port for computer (rear panel) USB type A port for printer (rear panel) |
|
Silicon Photodiode | |
Tungsten Halogen | |
100/240 V | |
0.002 A/hr. after warmup | |
9.8 in. | |
50/60 Hz | |
Sealed membrane with tactile response keys | |
<0.001 A at 0.0 A; <0.002 A at 2.0 A Peak-to peak at 340nm | |
Up to 3.0A at 340nm | |
External Printer: Ink Jet, Laser Printer with HP PCL 5 or Greater | |
<0.08 %T at 220 nm and 340 nm; <0.1 %T at 340 nm and 400 nm; <1.0 %T at 198 nm | |
1.0; 2.0; 3.0; 5.0 nm | |
11,000 nm/min. | |
8.6 kg | |
30 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.