Learn More
Innovative Research Inc Innovative Grade US Origin Horse Whole Blood Defibrinated 100ml
Supplier:  Innovative Research Inc IGHSWBDF100ML
Encompass_Preferred
Innovative Grade US Origin Horse Whole Blood Defibrinated from Innovative Research is collected per-order from horses in the United States. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used. The blood is mechanically defibrinated without anticoagulants or other additives. Our proprietary screening process ensures low fragility and consistent color. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4598