Learn More
Research Products International Corp Orbital and Shaking Water Bath
SDP
Supplier:  Research Products International Corp 246605
Encompass_Preferred
The Shaking Water Bath with Orbital and Reciprocating shaking is ideal for labs performing Northern and Southern hybridizations, gel de-staining, cell or bacterial culture incubation and washing procedures. The space saving design allows efficient use of bench space and features a high quality, durable design. Choose from a variety of trays to maximize your versatility. PID Controller allows easy temperature selection, rapid heat up, excellent stability and 3 point factory programmed temperature calibration ensures consistent temperature control. Unique magnetically coupled shaking mechanism offers maximum reliability, consistency and quiet operation. Easy to use - Switch from orbital to reciprocating mode simply by reversing the tray. Convenient drain port with quick disconnect allows easy cleaning. Optional trays include Universal Tray, Test Tube Tray and Perforated Tray. Polypropylene Spheres can be used as an alternative to a lid.
Catalog No. 50-222-9530